Product Details

SNP ID
rs150202548
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:170991458 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCACTGGCATGTTATTCAAAATCT[A/G]TAACATTTCTTTGGGAAACTTTTAG
Phenotype
MIM: 608438
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TLK1 PubMed Links

Gene Details

Gene
TLK1
Gene Name
tousled like kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136554.1 4654 UTR 3 NP_001130026.1
NM_001136555.1 4654 UTR 3 NP_001130027.1
NM_012290.4 4654 UTR 3 NP_036422.3
XM_011512238.2 4654 Intron XP_011510540.1
XM_017005421.1 4654 Intron XP_016860910.1
XM_017005422.1 4654 Intron XP_016860911.1

View Full Product Details