Product Details

SNP ID
rs150493401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:73701066 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATGTCTACATACCGCGTCCAGGG[A/T]TTTTTGGCAAGGAACCACAGGGCAG
Phenotype
MIM: 608190
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NAT8B PubMed Links

Gene Details

Gene
NAT8B
Gene Name
N-acetyltransferase 8B (putative, gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016347.2 275 Missense Mutation AAA,AAT K80N NP_057431.2

View Full Product Details