Product Details

SNP ID
rs151283587
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:206176529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTGTCCAGAAATGCCTACTGGAG[A/G]TCAGGATGCTTCGCTTCTTCACCTT
Phenotype
MIM: 600239
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR1 PubMed Links

Gene Details

Gene
GPR1
Gene Name
G protein-coupled receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098199.1 943 Missense Mutation ACC,ATC T240I NP_001091669.1
NM_001261452.1 943 Missense Mutation ACC,ATC T240I NP_001248381.1
NM_001261453.1 943 Missense Mutation ACC,ATC T240I NP_001248382.1
NM_001261454.1 943 Missense Mutation ACC,ATC T240I NP_001248383.1
NM_001261455.1 943 Missense Mutation ACC,ATC T240I NP_001248384.1
NM_005279.3 943 Missense Mutation ACC,ATC T240I NP_005270.2
XM_005246471.2 943 Missense Mutation ACC,ATC T240I XP_005246528.1
XM_017003832.1 943 Missense Mutation ACC,ATC T240I XP_016859321.1

View Full Product Details