Product Details
- SNP ID
-
rs138022720
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:119500728 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAGAATTTCAAAGGACCTTGCTAA[C/T]ATCTGTAAGACGGCAGCTACAGCAG
- Phenotype
-
MIM: 615618
MIM: 615534
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
POGLUT1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11539377] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- POGLUT1
- Gene Name
- protein O-glucosyltransferase 1
There are no transcripts associated with this gene.
- Gene
- TIMMDC1
- Gene Name
- translocase of inner mitochondrial membrane domain containing 1
View Full Product Details