Product Details

SNP ID
rs138023136
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:57557078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACTGAGACTGATGTGGAGGAGCG[A/T]GTCTACACCCCGTCCAATGCCGACA
Phenotype
MIM: 616519
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PDE12 PubMed Links

Gene Details

Gene
PDE12
Gene Name
phosphodiesterase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322176.1 832 Silent Mutation CGA,CGT R233R NP_001309105.1
NM_001322177.1 832 Silent Mutation CGA,CGT R233R NP_001309106.1
NM_177966.6 832 Silent Mutation CGA,CGT R233R NP_808881.3

View Full Product Details