Product Details

SNP ID
rs138103784
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:158646854 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGCCAGACCATGACTGTCAATC[A/G]TCAGATGAAGCGCTACAACGTTCCG
Phenotype
MIM: 606639
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GFM1 PubMed Links

Gene Details

Gene
GFM1
Gene Name
G elongation factor mitochondrial 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308164.1 617 Missense Mutation CAT,CGT H160R NP_001295093.1
NM_001308166.1 617 Missense Mutation CAT,CGT H160R NP_001295095.1
NM_024996.5 617 Missense Mutation CAT,CGT H160R NP_079272.4
XM_006713795.1 617 Missense Mutation CAT,CGT H160R XP_006713858.1

View Full Product Details