Product Details

SNP ID
rs138328955
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:14682757 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCTAGAATTATATCAGCAATACA[C/T]AGCCATGGCCCCCAAGCTACTGGCC
Phenotype
MIM: 616735
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C3orf20 PubMed Links

Gene Details

Gene
C3orf20
Gene Name
chromosome 3 open reading frame 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184957.1 496 Intron NP_001171886.1
NM_001184958.1 496 Intron NP_001171887.1
NM_032137.4 496 Missense Mutation ACA,ATA T15I NP_115513.4
XM_006713341.2 496 Missense Mutation ACA,ATA T15I XP_006713404.1
XM_006713342.2 496 Missense Mutation ACA,ATA T15I XP_006713405.1
XM_006713343.2 496 Missense Mutation ACA,ATA T15I XP_006713406.1
XM_011534153.2 496 Missense Mutation ACA,ATA T15I XP_011532455.1
XM_011534154.2 496 Intron XP_011532456.1
XM_011534157.2 496 Intron XP_011532459.1
XM_011534158.2 496 Missense Mutation ACA,ATA T15I XP_011532460.1
XM_017007310.1 496 Missense Mutation ACA,ATA T15I XP_016862799.1
Gene
CCDC174
Gene Name
coiled-coil domain containing 174
There are no transcripts associated with this gene.

View Full Product Details