Product Details

SNP ID
rs138358071
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:24120779 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAACAATCTGTCTGTAGATAAAGC[A/C]GGTAATCTGCCAATGAATTTCGTGC
Phenotype
MIM: 190160
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
THRB PubMed Links

Gene Details

Gene
THRB
Gene Name
thyroid hormone receptor beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000461.4 3913 UTR 3 NP_000452.2
NM_001128176.2 3913 UTR 3 NP_001121648.1
NM_001128177.1 3913 UTR 3 NP_001121649.1
NM_001252634.1 3913 UTR 3 NP_001239563.1
XM_005265421.4 3913 UTR 3 XP_005265478.1
XM_005265423.4 3913 UTR 3 XP_005265480.1
XM_005265424.3 3913 UTR 3 XP_005265481.1
XM_006713317.3 3913 UTR 3 XP_006713380.1
XM_006713318.3 3913 UTR 3 XP_006713381.1
XM_011534046.2 3913 UTR 3 XP_011532348.1
XM_011534047.2 3913 UTR 3 XP_011532349.1
XM_011534048.2 3913 UTR 3 XP_011532350.1
XM_011534049.2 3913 UTR 3 XP_011532351.1
XM_011534050.2 3913 UTR 3 XP_011532352.1
XM_011534051.2 3913 UTR 3 XP_011532353.1
XM_011534052.2 3913 UTR 3 XP_011532354.1
XM_011534053.2 3913 UTR 3 XP_011532355.1
XM_017007108.1 3913 UTR 3 XP_016862597.1
XM_017007109.1 3913 UTR 3 XP_016862598.1
XM_017007110.1 3913 UTR 3 XP_016862599.1
XM_017007111.1 3913 UTR 3 XP_016862600.1
XM_017007112.1 3913 UTR 3 XP_016862601.1
XM_017007113.1 3913 UTR 3 XP_016862602.1
XM_017007114.1 3913 UTR 3 XP_016862603.1
XM_017007115.1 3913 UTR 3 XP_016862604.1

View Full Product Details