Product Details

SNP ID
rs138782540
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:179329621 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATTTTTCATTTTATGTTCAGATT[A/C]CTCTGGAATAAGCAGAATTGCAGAT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF639 PubMed Links

Gene Details

Gene
ZNF639
Gene Name
zinc finger protein 639
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303425.1 807 Missense Mutation TAC,TCC Y21S NP_001290354.1
NM_001303426.1 807 Missense Mutation TAC,TCC Y21S NP_001290355.1
NM_016331.2 807 Missense Mutation TAC,TCC Y21S NP_057415.1
XM_017006550.1 807 Missense Mutation TAC,TCC Y21S XP_016862039.1
XM_017006551.1 807 Missense Mutation TAC,TCC Y21S XP_016862040.1
XM_017006552.1 807 Missense Mutation TAC,TCC Y21S XP_016862041.1
XM_017006553.1 807 Missense Mutation TAC,TCC Y21S XP_016862042.1

View Full Product Details