Product Details

SNP ID
rs139090920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:160677354 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGAGAAAAAGGCTGAAATGGGTT[C/T]GCTGGGTTCTAAAAATCCGCAAACC
Phenotype
MIM: 614439
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARL14 PubMed Links

Gene Details

Gene
ARL14
Gene Name
ADP ribosylation factor like GTPase 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025047.2 195 Missense Mutation TCG,TTG S3L NP_079323.1

View Full Product Details