Product Details

SNP ID
rs139593995
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:44658778 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGCTGAAAAACCCCTCATCATA[C/T]CAGAAAGAATCCAGAAAGCTGATCC
Phenotype
MIM: 194533
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF35 PubMed Links

Gene Details

Gene
ZNF35
Gene Name
zinc finger protein 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003420.3 Intron NP_003411.3
XM_017007129.1 Intron XP_016862618.1
XM_017007130.1 Intron XP_016862619.1
XM_017007131.1 Intron XP_016862620.1

View Full Product Details