Product Details

SNP ID
rs139668217
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:101522993 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGTGCTTAAACTACTTTTGTCA[C/T]CTAAGATTTTTTTGTCCTTCTGCAA
Phenotype
MIM: 612846
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM172BP PubMed Links

Gene Details

Gene
FAM172BP
Gene Name
family with sequence similarity 172, member B pseudogene
There are no transcripts associated with this gene.

Gene
SENP7
Gene Name
SUMO1/sentrin specific peptidase 7
There are no transcripts associated with this gene.

View Full Product Details