Product Details

SNP ID
rs141093031
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52247016 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGCACTGTTCGATGGGCACGGC[A/G]GTCCTGCAGCAGCCATCTTGGCTGC
Phenotype
MIM: 608979 MIM: 607433 MIM: 611059
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101929054 PubMed Links

Gene Details

Gene
LOC101929054
Gene Name
uncharacterized LOC101929054
There are no transcripts associated with this gene.

Gene
PPM1M
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122870.2 306 Intron NP_001116342.1
NM_144641.3 306 Missense Mutation AGT,GGT S129G NP_653242.3
XM_005264879.2 306 Missense Mutation AGT,GGT S129G XP_005264936.1
XM_005264880.3 306 Intron XP_005264937.1
XM_005264881.1 306 UTR 5 XP_005264938.1
Gene
TWF2
Gene Name
twinfilin actin binding protein 2
There are no transcripts associated with this gene.

Gene
WDR82
Gene Name
WD repeat domain 82
There are no transcripts associated with this gene.

View Full Product Details