Product Details

SNP ID
rs141102051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:186613340 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCTTGAACCAGATTGATGAAGTA[A/G]AGGTGTGGCCTCAGGTAAGTGGACC
Phenotype
MIM: 138680
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AHSG PubMed Links

Gene Details

Gene
AHSG
Gene Name
alpha 2-HS glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001622.2 413 Missense Mutation AAG,GAG K67E NP_001613.2
XM_017005840.1 413 Missense Mutation AAG,GAG K67E XP_016861329.1

View Full Product Details