Product Details

SNP ID
rs141208672
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:93893080 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGATAGATTCTGCGTACAGTATCAC[A/G]CCTTCTGAATCATATGTCCGGAAAT
Phenotype
MIM: 176880
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PROS1 PubMed Links

Gene Details

Gene
PROS1
Gene Name
protein S (alpha)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000313.3 1436 Silent Mutation GGC,GGT G336G NP_000304.2
NM_001314077.1 1436 Silent Mutation GGC,GGT G368G NP_001301006.1

View Full Product Details