Product Details

SNP ID
rs141310936
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:51971649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGCACCTGGATGGTGCCCTCGC[A/G]CTGCTCCACGCTTGCTGCCATGCCT
Phenotype
MIM: 608503
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABHD14A PubMed Links

Gene Details

Gene
ABHD14A
Gene Name
abhydrolase domain containing 14A
There are no transcripts associated with this gene.

Gene
ABHD14A-ACY1
Gene Name
ABHD14A-ACY1 readthrough
There are no transcripts associated with this gene.

Gene
ABHD14B
Gene Name
abhydrolase domain containing 14B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146314.1 716 Missense Mutation CGC,TGC R8C NP_001139786.1
NM_001254753.1 716 Intron NP_001241682.1
NM_032750.2 716 Missense Mutation CGC,TGC R8C NP_116139.1
Gene
PCBP4
Gene Name
poly(rC) binding protein 4
There are no transcripts associated with this gene.

View Full Product Details