Product Details

SNP ID
rs141351060
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:14180630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATATTAATATTTTCTGTTTTCCTG[G/T]CACTGTTTTTAAATGCATTAACTAG
Phenotype
MIM: 607283 MIM: 613208
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LSM3 PubMed Links

Gene Details

Gene
LSM3
Gene Name
LSM3 homolog, U6 small nuclear RNA and mRNA degradation associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014463.2 Intron NP_055278.1
Gene
XPC
Gene Name
XPC complex subunit, DNA damage recognition and repair factor
There are no transcripts associated with this gene.

View Full Product Details