Product Details

SNP ID
rs141456042
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:57272235 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTATGGCAATGAAGTCAGTAGAT[A/G]GGAAACCAGTTATTCCTTCTACCTT
Phenotype
MIM: 604299
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
APPL1 PubMed Links
Additional Information
For this assay, SNP(s) [rs10670620] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
APPL1
Gene Name
adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012096.2 4878 UTR 3 NP_036228.1
XM_011533583.2 4878 UTR 3 XP_011531885.1
Gene
ASB14
Gene Name
ankyrin repeat and SOCS box containing 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142733.2 4878 Intron NP_001136205.2
NM_130387.5 4878 Intron NP_569058.1
XM_017005736.1 4878 Intron XP_016861225.1
XM_017005737.1 4878 Intron XP_016861226.1
Gene
LOC105377102
Gene Name
uncharacterized LOC105377102
There are no transcripts associated with this gene.

View Full Product Details