Product Details

SNP ID
rs141536671
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:114128799 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCACAGGGTTGAGGGCGCTATTCA[C/T]GTAGCCCAGCCATGTCGTGGCACTG
Phenotype
MIM: 126451
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DRD3 PubMed Links

Gene Details

Gene
DRD3
Gene Name
dopamine receptor D3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000796.5 1543 Missense Mutation NP_000787.2
NM_001282563.2 1543 Missense Mutation NP_001269492.1
NM_001290809.1 1543 Missense Mutation NP_001277738.1
NM_033663.5 1543 Missense Mutation NP_387512.3
XM_017005829.1 1543 Missense Mutation XP_016861318.1

View Full Product Details