Product Details

SNP ID
rs141729450
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:98516709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGTGTACTCTTTCCGGTTGGTGT[A/G]AGCAGCCCAGATGAAGAGAGCAGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLDND1 PubMed Links

Gene Details

Gene
CLDND1
Gene Name
claudin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040181.1 638 Missense Mutation CAC,TAC H238Y NP_001035271.1
NM_001040182.1 638 Missense Mutation CAC,TAC H261Y NP_001035272.1
NM_001040183.1 638 Missense Mutation CAC,TAC H238Y NP_001035273.1
NM_001040199.1 638 Missense Mutation CAC,TAC H238Y NP_001035289.1
NM_001040200.1 638 Missense Mutation CAC,TAC H143Y NP_001035290.1
NM_019895.2 638 Missense Mutation CAC,TAC H238Y NP_063948.1

View Full Product Details