Product Details

SNP ID
rs142390372
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:15073797 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTGCTGGTCAGTCAGTGCCCCC[A/C]TTCTGCTTGGCCAGGGTGTGCTTCA
Phenotype
MIM: 611987 MIM: 609511
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MRPS25 PubMed Links

Gene Details

Gene
MRPS25
Gene Name
mitochondrial ribosomal protein S25
There are no transcripts associated with this gene.

Gene
RBSN
Gene Name
rabenosyn, RAB effector
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302378.1 2799 Missense Mutation NP_001289307.1
NM_022340.3 2799 Missense Mutation NP_071735.2
XM_005265384.4 2799 Missense Mutation XP_005265441.1
XM_005265385.4 2799 Missense Mutation XP_005265442.1
XM_011534001.2 2799 Missense Mutation XP_011532303.1
XM_017007023.1 2799 Missense Mutation XP_016862512.1
XM_017007024.1 2799 Missense Mutation XP_016862513.1
XM_017007025.1 2799 Missense Mutation XP_016862514.1
XM_017007026.1 2799 Missense Mutation XP_016862515.1
XM_017007027.1 2799 Missense Mutation XP_016862516.1
XM_017007028.1 2799 Missense Mutation XP_016862517.1

View Full Product Details