Product Details

SNP ID
rs142768626
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:120325124 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAAAAACCTCAACTGCAGGCACA[C/T]GTCTTTACATGATTGTTGTAAAGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC58 PubMed Links

Gene Details

Gene
LRRC58
Gene Name
leucine rich repeat containing 58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099678.1 7288 UTR 3 NP_001093148.1

View Full Product Details