Product Details

SNP ID
rs143082277
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:185589418 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTTTGGGGATCATTTTCAAAAG[A/G]AAATTGCTTGTTTGTTGTTTTTAAA
Phenotype
MIM: 608261
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SENP2 PubMed Links

Gene Details

Gene
SENP2
Gene Name
SUMO1/sentrin/SMT3 specific peptidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021627.2 Intron NP_067640.2
XM_005247690.2 Intron XP_005247747.2
XM_005247691.2 Intron XP_005247748.1
XM_017006973.1 Intron XP_016862462.1

View Full Product Details