Product Details
- SNP ID
-
rs143372604
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:27114272 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTCTTTCTGTTCCTGTTACCATT[C/T]TCTTTTCCTAATGCATATACTCAGC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NEK10
PubMed Links
Gene Details
- Gene
- NEK10
- Gene Name
- NIMA related kinase 10
View Full Product Details