Product Details

SNP ID
rs143528366
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:44594362 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTATGTATGTACTGAGTGTGGG[A/C]AAGCCTTTAGTCAGAGTGCAAACCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF660 PubMed Links

Gene Details

Gene
ZNF660
Gene Name
zinc finger protein 660
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173658.2 502 Missense Mutation AAA,CAA K57Q NP_775929.2
XM_005265090.4 502 Missense Mutation AAA,CAA K57Q XP_005265147.1
XM_006713117.3 502 Missense Mutation AAA,CAA K57Q XP_006713180.1
XM_011533622.2 502 Missense Mutation AAA,CAA K57Q XP_011531924.1

View Full Product Details