Product Details

SNP ID
rs143795787
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:126482452 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGTCTGGCAGATGATCTCATAGG[C/T]CTTCTGGTTCCCTGACCAGCAGCGC
Phenotype
MIM: 613012 MIM: 615746
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UROC1 PubMed Links

Gene Details

Gene
UROC1
Gene Name
urocanate hydratase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165974.1 1978 Missense Mutation ACC,GCC T702A NP_001159446.1
NM_144639.2 1978 Missense Mutation ACC,GCC T642A NP_653240.1
Gene
ZXDC
Gene Name
ZXD family zinc finger C
There are no transcripts associated with this gene.

View Full Product Details