Product Details

SNP ID
rs143982217
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:119500785 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGGCTGGGTGTATGGGGGAATACC[A/G]GCTTTTATTCATGCTAAACAACAAT
Phenotype
MIM: 615618 MIM: 615534
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POGLUT1 PubMed Links

Gene Details

Gene
POGLUT1
Gene Name
protein O-glucosyltransferase 1
There are no transcripts associated with this gene.

Gene
TIMMDC1
Gene Name
translocase of inner mitochondrial membrane domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016589.3 498 Silent Mutation CCA,CCG P95P NP_057673.2
XM_017006556.1 498 Intron XP_016862045.1

View Full Product Details