Product Details

SNP ID
rs144011121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:160677355 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGAAAAAGGCTGAAATGGGTTC[A/G]CTGGGTTCTAAAAATCCGCAAACCA
Phenotype
MIM: 614439
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARL14 PubMed Links

Gene Details

Gene
ARL14
Gene Name
ADP ribosylation factor like GTPase 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025047.2 196 Silent Mutation TCA,TCG S3S NP_079323.1

View Full Product Details