Product Details

SNP ID
rs144124655
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:172506868 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCAGGAATGAATGCCCACTCCTT[A/G]ATGATTCCCAGGAGTTTATTTTGCG
Phenotype
MIM: 603598
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNFSF10 PubMed Links

Gene Details

Gene
TNFSF10
Gene Name
tumor necrosis factor superfamily member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190942.1 593 UTR 3 NP_001177871.1
NM_001190943.1 593 Intron NP_001177872.1
NM_003810.3 593 Missense Mutation TCA,TTA S157L NP_003801.1

View Full Product Details