Product Details
- SNP ID
-
rs144176995
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:131029796 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAGACCTTGATTGCAAGAAGATAC[A/G]TGCTTCAACAAAAACTTGGCAGTGG
- Phenotype
-
MIM: 609779
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ASTE1
PubMed Links
Gene Details
- Gene
- ASTE1
- Gene Name
- asteroid homolog 1 (Drosophila)
There are no transcripts associated with this gene.
- Gene
- NEK11
- Gene Name
- NIMA related kinase 11
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001146003.1 |
516 |
Missense Mutation |
ATG,GTG |
M30V |
NP_001139475.1 |
NM_001321220.1 |
516 |
Missense Mutation |
ATG,GTG |
M30V |
NP_001308149.1 |
NM_001321221.1 |
516 |
Missense Mutation |
ATG,GTG |
M30V |
NP_001308150.1 |
NM_001321222.1 |
516 |
Missense Mutation |
ATG,GTG |
M30V |
NP_001308151.1 |
NM_001321223.1 |
516 |
Missense Mutation |
ATG,GTG |
M30V |
NP_001308152.1 |
NM_001321224.1 |
516 |
UTR 5 |
|
|
NP_001308153.1 |
NM_024800.4 |
516 |
Missense Mutation |
ATG,GTG |
M30V |
NP_079076.3 |
NM_145910.3 |
516 |
Missense Mutation |
ATG,GTG |
M30V |
NP_665917.1 |
XM_011513167.2 |
516 |
Missense Mutation |
ATG,GTG |
M30V |
XP_011511469.1 |
XM_011513173.1 |
516 |
UTR 5 |
|
|
XP_011511475.1 |
XM_011513174.1 |
516 |
Intron |
|
|
XP_011511476.1 |
XM_011513175.1 |
516 |
Intron |
|
|
XP_011511477.1 |
XM_011513181.1 |
516 |
Intron |
|
|
XP_011511483.1 |
XM_017007205.1 |
516 |
Missense Mutation |
ATG,GTG |
M30V |
XP_016862694.1 |
XM_017007206.1 |
516 |
Missense Mutation |
ATG,GTG |
M30V |
XP_016862695.1 |
XM_017007207.1 |
516 |
Missense Mutation |
ATG,GTG |
M30V |
XP_016862696.1 |
XM_017007208.1 |
516 |
Missense Mutation |
ATG,GTG |
M30V |
XP_016862697.1 |
XM_017007209.1 |
516 |
Missense Mutation |
ATG,GTG |
M30V |
XP_016862698.1 |
XM_017007210.1 |
516 |
Missense Mutation |
ATG,GTG |
M30V |
XP_016862699.1 |
XM_017007211.1 |
516 |
Missense Mutation |
ATG,GTG |
M30V |
XP_016862700.1 |
XM_017007212.1 |
516 |
Missense Mutation |
ATG,GTG |
M30V |
XP_016862701.1 |
XM_017007213.1 |
516 |
Missense Mutation |
ATG,GTG |
M30V |
XP_016862702.1 |
XM_017007214.1 |
516 |
Missense Mutation |
ATG,GTG |
M30V |
XP_016862703.1 |
XM_017007215.1 |
516 |
Missense Mutation |
ATG,GTG |
M30V |
XP_016862704.1 |
XM_017007216.1 |
516 |
Missense Mutation |
ATG,GTG |
M30V |
XP_016862705.1 |
XM_017007217.1 |
516 |
Missense Mutation |
ATG,GTG |
M30V |
XP_016862706.1 |
XM_017007218.1 |
516 |
UTR 5 |
|
|
XP_016862707.1 |
XM_017007219.1 |
516 |
Intron |
|
|
XP_016862708.1 |
XM_017007220.1 |
516 |
Missense Mutation |
ATG,GTG |
M30V |
XP_016862709.1 |
XM_017007221.1 |
516 |
Missense Mutation |
ATG,GTG |
M30V |
XP_016862710.1 |
XM_017007222.1 |
516 |
Missense Mutation |
ATG,GTG |
M30V |
XP_016862711.1 |
XM_017007223.1 |
516 |
Missense Mutation |
ATG,GTG |
M30V |
XP_016862712.1 |
XM_017007224.1 |
516 |
UTR 5 |
|
|
XP_016862713.1 |
XM_017007225.1 |
516 |
UTR 5 |
|
|
XP_016862714.1 |
XM_017007226.1 |
516 |
UTR 5 |
|
|
XP_016862715.1 |
XM_017007227.1 |
516 |
UTR 5 |
|
|
XP_016862716.1 |
XM_017007228.1 |
516 |
Intron |
|
|
XP_016862717.1 |
XM_017007229.1 |
516 |
Missense Mutation |
ATG,GTG |
M30V |
XP_016862718.1 |
XM_017007230.1 |
516 |
Missense Mutation |
ATG,GTG |
M30V |
XP_016862719.1 |
XM_017007231.1 |
516 |
UTR 5 |
|
|
XP_016862720.1 |
XM_017007232.1 |
516 |
UTR 5 |
|
|
XP_016862721.1 |
XM_017007233.1 |
516 |
UTR 5 |
|
|
XP_016862722.1 |
View Full Product Details