Product Details

SNP ID
rs145066209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52451521 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCATCCAGGTCGATGTAGCCATC[A/G]GCATTTCTGTGGGGAGGGGGCTCAG
Phenotype
MIM: 615507 MIM: 191040
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NISCH PubMed Links

Gene Details

Gene
NISCH
Gene Name
nischarin
There are no transcripts associated with this gene.

Gene
SEMA3G
Gene Name
semaphorin 3G
There are no transcripts associated with this gene.

Gene
TNNC1
Gene Name
troponin C1, slow skeletal and cardiac type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003280.2 350 Silent Mutation GCC,GCT A108A NP_003271.1

View Full Product Details