Product Details

SNP ID
rs145152566
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:160508174 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCAGCCATTTTTAATATATTACT[C/T]AGTCCATCGAGTACTACTTGCACAA
Phenotype
MIM: 602970 MIM: 615644
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
KPNA4 PubMed Links

Gene Details

Gene
KPNA4
Gene Name
karyopherin subunit alpha 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002268.4 1611 Silent Mutation CTA,CTG L435L NP_002259.1
Gene
SCARNA7
Gene Name
small Cajal body-specific RNA 7
There are no transcripts associated with this gene.

View Full Product Details