Product Details

SNP ID
rs145240396
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196306988 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCCAATCACTACTTGCACCACC[A/G]TTTTGTATCGGTCAAATCCCATTTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TCTEX1D2 PubMed Links

Gene Details

Gene
TCTEX1D2
Gene Name
Tctex1 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152773.4 414 Missense Mutation ACG,ATG T91M NP_689986.2
Gene
TM4SF19-TCTEX1D2
Gene Name
TM4SF19-TCTEX1D2 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details