Product Details

SNP ID
rs145301682
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:133400238 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAATGCCATGAGTGGCCTTGTCC[A/G]AGCACCCGGGGTCTATGTAGGAACA
Phenotype
MIM: 603212
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BFSP2 PubMed Links

Gene Details

Gene
BFSP2
Gene Name
beaded filament structural protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003571.3 2357 Missense Mutation CAA,CGA Q52R NP_003562.1
XM_017007315.1 2357 Missense Mutation CAA,CGA Q52R XP_016862804.1
Gene
TMEM108
Gene Name
transmembrane protein 108
There are no transcripts associated with this gene.

View Full Product Details