Product Details

SNP ID
rs145417442
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:161224985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAAATCCTGCCAATATTTGTGTG[A/G]CCTGTTTGCGAAGTAAAGTGGACAT
Phenotype
MIM: 611021
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NMD3 PubMed Links

Gene Details

Gene
NMD3
Gene Name
NMD3 ribosome export adaptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320227.1 240 Missense Mutation ACC,GCC T34A NP_001307156.1
NM_015938.4 240 Intron NP_057022.2
XM_005247511.1 240 Missense Mutation ACC,GCC T34A XP_005247568.1
XM_005247512.1 240 Missense Mutation ACC,GCC T34A XP_005247569.1

View Full Product Details