Product Details
- SNP ID
-
rs145440461
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:129383313 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAATCAGTACAATATGATGTTTT[G/T]AAAAAGCTGGATGATTTTGACAATA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RPL32P3
PubMed Links
Gene Details
- Gene
- RPL32P3
- Gene Name
- ribosomal protein L32 pseudogene 3
There are no transcripts associated with this gene.
View Full Product Details