Product Details

SNP ID
rs145536359
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:172506584 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGAAACAAAAATTCTGTCATTTT[C/T]CTTAAGCTCAAATATTCCCCCTTGA
Phenotype
MIM: 603598
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TNFSF10 PubMed Links

Gene Details

Gene
TNFSF10
Gene Name
tumor necrosis factor superfamily member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190942.1 877 UTR 3 NP_001177871.1
NM_001190943.1 877 Intron NP_001177872.1
NM_003810.3 877 Missense Mutation AAA,GAA K252E NP_003801.1

View Full Product Details