Product Details

SNP ID
rs146071566
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:136151806 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGTGTTGGGCCTCGGATAATGG[C/T]AGAAATTGGAAGTGGCTGAACTTTC
Phenotype
MIM: 614802 MIM: 604944
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MSL2 PubMed Links

Gene Details

Gene
MSL2
Gene Name
male-specific lethal 2 homolog (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145417.1 1808 Missense Mutation ACC,GCC T285A NP_001138889.1
NM_018133.3 1808 Missense Mutation ACC,GCC T359A NP_060603.2
XM_005247571.3 1808 Missense Mutation ACC,GCC T285A XP_005247628.1
XM_006713684.3 1808 Missense Mutation ACC,GCC T285A XP_006713747.1
XM_011512949.2 1808 Missense Mutation ACC,GCC T285A XP_011511251.1
Gene
PPP2R3A
Gene Name
protein phosphatase 2 regulatory subunit B''alpha
There are no transcripts associated with this gene.

View Full Product Details