Product Details

SNP ID
rs146412696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:15170334 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCTTCGTCCTTCCTTTTCTTTCC[C/T]GGTAGGTTAATTTGTCCAAATGCTC
Phenotype
MIM: 612397
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COL6A4P1 PubMed Links

Gene Details

Gene
COL6A4P1
Gene Name
collagen type VI alpha 4 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details