Product Details

SNP ID
rs146651021
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196935933 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGGTAAAATTTGAGGAAAAAAAT[A/G]AGAAATGGCTAAAAGAGGCATCTGC
Phenotype
MIM: 605133 MIM: 612845
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NCBP2 PubMed Links

Gene Details

Gene
NCBP2
Gene Name
nuclear cap binding protein subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042540.1 1480 UTR 3 NP_001036005.1
NM_001308036.1 1480 UTR 3 NP_001294965.1
NM_007362.3 1480 UTR 3 NP_031388.2
XM_011512556.2 1480 UTR 3 XP_011510858.1
XM_011512557.2 1480 UTR 3 XP_011510859.1
XM_011512558.2 1480 UTR 3 XP_011510860.1
Gene
NCBP2-AS2
Gene Name
NCBP2 antisense RNA 2 (head to head)
There are no transcripts associated with this gene.

Gene
SENP5
Gene Name
SUMO1/sentrin specific peptidase 5
There are no transcripts associated with this gene.

View Full Product Details