Product Details

SNP ID
rs146737733
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:8880079 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTAGTTTCCTTAATGCACCCCTC[A/T]GGATTTCATGCAAATGAAATTATTA
Phenotype
MIM: 605256
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RAD18 PubMed Links

Gene Details

Gene
RAD18
Gene Name
RAD18, E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020165.3 2856 UTR 3 NP_064550.3
XM_017006873.1 2856 Intron XP_016862362.1

View Full Product Details