Product Details
- SNP ID
-
rs147014655
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:50572194 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATGGTGCCCCCAGTGTTTATTCCT[C/T]GGCCAGAAACAGAGGTAGGTGTGCC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C3orf18
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2232248] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C3orf18
- Gene Name
- chromosome 3 open reading frame 18
- Gene
- HEMK1
- Gene Name
- HemK methyltransferase family member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001317851.1 |
997 |
Missense Mutation |
CGG,TGG |
R134W |
NP_001304780.1 |
NM_016173.4 |
997 |
Missense Mutation |
CGG,TGG |
R134W |
NP_057257.1 |
XM_005265218.3 |
997 |
Missense Mutation |
CGG,TGG |
R134W |
XP_005265275.1 |
XM_011533806.2 |
997 |
Missense Mutation |
CGG,TGG |
R134W |
XP_011532108.1 |
XM_011533807.1 |
997 |
Missense Mutation |
CGG,TGG |
R134W |
XP_011532109.1 |
XM_011533808.1 |
997 |
Missense Mutation |
CGG,TGG |
R134W |
XP_011532110.1 |
XM_011533809.2 |
997 |
Missense Mutation |
CGG,TGG |
R134W |
XP_011532111.1 |
XM_011533810.2 |
997 |
Missense Mutation |
CGG,TGG |
R134W |
XP_011532112.1 |
XM_011533812.2 |
997 |
Missense Mutation |
CGG,TGG |
R134W |
XP_011532114.1 |
XM_011533813.2 |
997 |
Missense Mutation |
CGG,TGG |
R27W |
XP_011532115.1 |
XM_011533814.2 |
997 |
Missense Mutation |
CGG,TGG |
R27W |
XP_011532116.1 |
View Full Product Details