Product Details
- SNP ID
-
rs147125759
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:46859491 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCCCTCACCCAGCGTGGCCAGCAC[A/G]TGGCGAAGCTCAGCACCCATGACAG
- Phenotype
-
MIM: 160790
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MYL3
PubMed Links
Gene Details
- Gene
- MYL3
- Gene Name
- myosin light chain 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000258.2 |
558 |
Silent Mutation |
CAC,CAT |
H155H |
NP_000249.1 |
View Full Product Details