Product Details

SNP ID
rs147377926
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:47030363 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAAATCAGTGAACTTTAAATAGA[A/G]GCCAACAGAAACCACGCAAACTGAA
Phenotype
MIM: 612778
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SETD2 PubMed Links

Gene Details

Gene
SETD2
Gene Name
SET domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014159.6 Intron NP_054878.5
XM_011533631.2 Intron XP_011531933.1
XM_011533632.2 Intron XP_011531934.1
XM_011533633.2 Intron XP_011531935.1
XM_011533634.1 Intron XP_011531936.1
XM_017006270.1 Intron XP_016861759.1

View Full Product Details