Product Details

SNP ID
rs147759114
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:158669097 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCAACCCAGGCTAAATGTAGAA[C/T]GAGCGTCATTTCTGGAGATACATTT
Phenotype
MIM: 606639 MIM: 609305
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GFM1 PubMed Links

Gene Details

Gene
GFM1
Gene Name
G elongation factor mitochondrial 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308164.1 650 Intron NP_001295093.1
NM_001308166.1 650 Intron NP_001295095.1
NM_024996.5 650 Intron NP_079272.4
XM_006713795.1 650 Intron XP_006713858.1
Gene
LXN
Gene Name
latexin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020169.3 650 Missense Mutation ATT,GTT I136V NP_064554.3
XM_017006876.1 650 Missense Mutation ATT,GTT I136V XP_016862365.1

View Full Product Details