Product Details

SNP ID
rs147857480
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:158669566 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACTTCTGGTGCAGTTTCTTGTCC[C/T]GTTGAAGGGTAAAGTACTTCAGCTG
Phenotype
MIM: 606639 MIM: 609305
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GFM1 PubMed Links

Gene Details

Gene
GFM1
Gene Name
G elongation factor mitochondrial 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308164.1 481 Intron NP_001295093.1
NM_001308166.1 481 Intron NP_001295095.1
NM_024996.5 481 Intron NP_079272.4
XM_006713795.1 481 Intron XP_006713858.1
Gene
LXN
Gene Name
latexin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020169.3 481 Silent Mutation ACA,ACG T79T NP_064554.3
XM_017006876.1 481 Silent Mutation ACA,ACG T79T XP_016862365.1

View Full Product Details