Product Details

SNP ID
rs148258453
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49887318 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCAAGAACTAAGTCAAGTGGGCC[A/G]AGGAGGCTCTGAGAGTGGCCGGGGC
Phenotype
MIM: 600168
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MST1R PubMed Links

Gene Details

Gene
MST1R
Gene Name
macrophage stimulating 1 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244937.2 4218 Missense Mutation CGG,TGG R1349W NP_001231866.1
NM_001318913.1 4218 Missense Mutation CGG,TGG R1292W NP_001305842.1
NM_002447.3 4218 Missense Mutation CGG,TGG R1398W NP_002438.2
XM_005265170.4 4218 Missense Mutation CGG,TGG R1399W XP_005265227.2
XM_011533739.2 4218 Missense Mutation CGG,TGG R1359W XP_011532041.1
XM_011533740.2 4218 Missense Mutation CGG,TGG R1350W XP_011532042.1
XM_011533741.2 4218 Missense Mutation CGG,TGG R1330W XP_011532043.1
XM_011533742.2 4218 UTR 3 XP_011532044.1
XM_011533743.2 4218 Missense Mutation CGG,TGG R1298W XP_011532045.1
XM_011533744.2 4218 Missense Mutation CGG,TGG R1293W XP_011532046.1

View Full Product Details