Product Details

SNP ID
rs148652006
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:185590162 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGATGAAATACCAGCCAAAAGACC[A/G]AGATTAGGTACTGAATATAAATTTT
Phenotype
MIM: 608261
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SENP2 PubMed Links

Gene Details

Gene
SENP2
Gene Name
SUMO1/sentrin/SMT3 specific peptidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021627.2 267 Silent Mutation CCA,CCG P50P NP_067640.2
XM_005247690.2 267 Silent Mutation CCA,CCG P50P XP_005247747.2
XM_005247691.2 267 UTR 5 XP_005247748.1
XM_017006973.1 267 UTR 5 XP_016862462.1

View Full Product Details