Product Details

SNP ID
rs148935732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:32953484 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAGCATATACAGCAATTACTATC[A/T]GTATGAAAGTATCCCCAAGCCTTGC
Phenotype
MIM: 604836
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CCR4 PubMed Links

Gene Details

Gene
CCR4
Gene Name
C-C motif chemokine receptor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005508.4 277 Missense Mutation CAG,CTG Q21L NP_005499.1
XM_017005687.1 277 Missense Mutation CAG,CTG Q21L XP_016861176.1
Gene
LOC105377022
Gene Name
uncharacterized LOC105377022
There are no transcripts associated with this gene.

View Full Product Details