Product Details

SNP ID
rs149073292
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50286785 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGCTGCAGCCCTGTGTACAGAC[A/G]AGGAGGGTTCCTGCTGCTGCTCGCG
Phenotype
MIM: 604038 MIM: 602725 MIM: 607073 MIM: 601281
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HYAL3 PubMed Links

Gene Details

Gene
HYAL3
Gene Name
hyaluronoglucosaminidase 3
There are no transcripts associated with this gene.

Gene
IFRD2
Gene Name
interferon related developmental regulator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006764.4 368 Intron NP_006755.4
Gene
LSMEM2
Gene Name
leucine rich single-pass membrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304385.1 368 Missense Mutation CAA,CGA Q94R NP_001291314.1
NM_153215.2 368 Missense Mutation CAA,CGA Q95R NP_694947.1
XM_006712979.3 368 Missense Mutation CAA,CGA Q104R XP_006713042.1
XM_006712980.3 368 Missense Mutation CAA,CGA Q103R XP_006713043.1
XM_011533370.2 368 Missense Mutation CAA,CGA Q71R XP_011531672.1
Gene
NAT6
Gene Name
N-acetyltransferase 6
There are no transcripts associated with this gene.

Gene
SEMA3B
Gene Name
semaphorin 3B
There are no transcripts associated with this gene.

View Full Product Details